1,3,4-oxadiazol-2-amine 95%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 1,3,4-Oxadiazol-2-amine

OR54600-1G
APOSOR54600-1GEA 79.9 EUR
APOSOR54600-1G
1,3,4-oxadiazol-2-amine 95%
1,3,4-oxadiazol-2-amine
Formel: C₂H₃N₃O
Molecular Weight: 85,065 g/mol
Schmelzpunkt: 153,3 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD17677516
CAS-Nummer: 3775-60-8
UN: 3259
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR