6-Bromobenzofuran
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
6-Bromo-1-benzofuran
OR54601-1G
OR54601-5G
APOSOR54601-1GEA
66.7
EUR
APOSOR54601-1G
APOSOR54601-5G
6-Bromobenzofuran
6-Bromobenzofuran
Formel:
C₈H₅BrO Molecular Weight: 197,029 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08669477 CAS-Nummer: 128851-73-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...