4-Bromobenzofuran

Lieferant: Apollo Scientific
Achtung

OR54602-1G OR54602-250MG
APOSOR54602-1GEA 229 EUR
APOSOR54602-1G APOSOR54602-250MG
4-Bromobenzofuran
4-Bromobenzofuran
Formel: C₈H₅BrO
Molecular Weight: 197,029 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09056755
CAS-Nummer: 128868-60-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR