Dimethyl 2-bromoisophthalate 95+%

Lieferant: Apollo Scientific

Synonyme: Dimethyl 2-bromobenzene-1,3-dicarboxylate, 2-bromoisophthalic acid methyl ester

OR54603-250MG OR54603-1G
APOSOR54603-250MGEA 82.9 EUR
APOSOR54603-250MG APOSOR54603-1G
Dimethyl 2-bromoisophthalate 95+%
Dimethyl 2-bromoisophthalate
Formel: C₁₀H₉BrO₄
Molecular Weight: 273.08 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD17169956
CAS-Nummer: 39622-80-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR