4-(Trifluoromethyl)thiazole-2-carboxaldehyde
Lieferant: Apollo Scientific
|
Achtung
|
OR54614-1G
OR54614-250MG
APOSOR54614-1GEA
1350
EUR
APOSOR54614-1G
APOSOR54614-250MG
4-(Trifluoromethyl)thiazole-2-carboxaldehyde
4-(Trifluoromethyl)thiazole-2-carboxaldehyde
Formel:
C₅H₂F₃NOS Lagertemperatur: Kühlschrank |
MDL:
MFCD09909625 CAS-Nummer: 354587-75-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...