4-Bromo-2-chloro-5-iodophenol 95%
Lieferant: Apollo Scientific
|
Gefahr
|
OR54637-1G
OR54637-250MG
APOSOR54637-1GEA
284
EUR
APOSOR54637-1G
APOSOR54637-250MG
4-Bromo-2-chloro-5-iodophenol 95%
4-Bromo-2-chloro-5-iodophenol
Formel:
C₆H₃BrClIO Molecular Weight: 333,349 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD31700577 CAS-Nummer: 2092799-35-2 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...