Methyl 4-aminooxane-4-carboxylate hydrochloride
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Methyl-4-aminotetrahydro-2H-pyran-4-carboxylate hydrochloride
OR54638-1G
OR54638-5G
APOSOR54638-1GEA
102
EUR
APOSOR54638-1G
APOSOR54638-5G
Methyl 4-aminooxane-4-carboxylate hydrochloride
Methyl 4-aminooxane-4-carboxylate hydrochloride
Formel:
C₇H₁₃NO₃·ClH Lagertemperatur: Raumtemperatur |
MDL:
MFCD11226845 CAS-Nummer: 199330-66-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...