4-Iodo-7-azaindoline 97%
Lieferant: Apollo Scientific
|
Achtung
|
OR54642-1G
OR54642-250MG
APOSOR54642-1GEA
0
EUR
APOSOR54642-1G
APOSOR54642-250MG
4-Iodo-7-azaindoline 97%
4-Iodo-7-azaindoline
Formel:
C₇H₇IN₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD22548136 CAS-Nummer: 945600-09-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...