2-(Cyclopropyl)benzyl bromide

Lieferant: Apollo Scientific
Gefahr

OR54649-1G OR54649-250MG
APOSOR54649-1GEA 551 EUR
APOSOR54649-1G APOSOR54649-250MG
2-(Cyclopropyl)benzyl bromide
2-(Cyclopropyl)benzyl bromide
Formel: C₁₀H₁₁Br
Molecular Weight: 211.098 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD15527184
CAS-Nummer: 1260801-90-8
UN: 1760
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR