2-(Cyclopropyl)benzyl alcohol

Lieferant: Apollo Scientific
Achtung

OR54651-1G OR54651-250MG OR54651-5G
APOSOR54651-1GEA 162 EUR
APOSOR54651-1G APOSOR54651-250MG APOSOR54651-5G
2-(Cyclopropyl)benzyl alcohol
2-(Cyclopropyl)benzyl alcohol
Formel: C₁₀H₁₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD20921818
CAS-Nummer: 118184-68-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR