7-Bromo-5-nitrobenzofuran-2-carboxylic acid
Lieferant: Apollo Scientific
|
Achtung
|
OR54655-1G
OR54655-250MG
APOSOR54655-1GEA
575
EUR
APOSOR54655-1G
APOSOR54655-250MG
7-Bromo-5-nitrobenzofuran-2-carboxylic acid
7-Bromo-5-nitrobenzofuran-2-carboxylic acid
Formel:
C₉H₄BrNO₅ Molecular Weight: 286,036 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD28098934 CAS-Nummer: 286836-15-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...