2-Hydroxy-3-cyclopropylbenzaldehyde
Lieferant: Apollo Scientific
|
Achtung
|
OR54661-1G
OR54661-250MG
APOSOR54661-1GEA
0
EUR
APOSOR54661-1G
APOSOR54661-250MG
2-Hydroxy-3-cyclopropylbenzaldehyde
2-Hydroxy-3-cyclopropylbenzaldehyde
Formel:
C₁₀H₁₀O₂ Lagertemperatur: Kühlschrank |
MDL:
MFCD31726535 CAS-Nummer: 1206776-33-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...