3,7-Dioxabicyclo[4.1.0]heptane 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4,7-Dioxabicyclo[4.1.0]heptane 95%

OR54673-1G OR54673-250MG
APOSOR54673-1GEA 111 EUR
APOSOR54673-1G APOSOR54673-250MG
3,7-Dioxabicyclo[4.1.0]heptane 95%
3,7-Dioxabicyclo[4.1.0]heptane
Formel: C₅H₈O₂
Molecular Weight: 100,116 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD11226853
CAS-Nummer: 286-22-6
UN: 1993
ADR: 3,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR