(2R)-4-Iodo-2-methyl-2,3-dihydro-1H-pyrrolo[2,3-b]pyridine 95%

Lieferant: Apollo Scientific
Achtung

OR54680-1G OR54680-250MG
APOSOR54680-1GEA 766 EUR
APOSOR54680-1G APOSOR54680-250MG
(2R)-4-Iodo-2-methyl-2,3-dihydro-1H-pyrrolo[2,3-b]pyridine 95%
(2R)-4-Iodo-2-methyl-2,3-dihydro-1H-pyrrolo[2,3-b]pyridine
Formel: C₈H₉IN₂
Molecular Weight: 260.075 g/mol
Lagertemperatur: Raumtemperatur
CAS-Nummer: 2366997-10-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR