6-Bromo-2-iodo-1H-indole 95%
Lieferant: Apollo Scientific
|
Achtung
|
OR54693-1G
OR54693-250MG
APOSOR54693-1GEA
654
EUR
APOSOR54693-1G
APOSOR54693-250MG
6-Bromo-2-iodo-1H-indole 95%
6-Bromo-2-iodo-1H-indole
Formel:
C₈H₅BrIN Lagertemperatur: Kühlschrank |
MDL:
MFCD28664103 CAS-Nummer: 1572177-89-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...