4-Bromo-2-iodo-1H-indole

Lieferant: Apollo Scientific
Achtung

OR54694-1G OR54694-250MG
APOSOR54694-1GEA 944 EUR
APOSOR54694-1G APOSOR54694-250MG
4-Bromo-2-iodo-1H-indole
4-Bromo-2-iodo-1H-indole
Formel: C₈H₅BrIN
Molecular Weight: 321,94 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD31915671
CAS-Nummer: 2309465-44-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR