5-Bromo-7-chloro-1-benzofuran-2-carboxylic acid

Lieferant: Apollo Scientific
Achtung

OR54701-1G OR54701-250MG
APOSOR54701-1GEA 434 EUR
APOSOR54701-1G APOSOR54701-250MG
5-Bromo-7-chloro-1-benzofuran-2-carboxylic acid
5-Bromo-7-chloro-1-benzofuran-2-carboxylic acid
Formel: C₉H₄BrClO₃
Molecular Weight: 275,483 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07429304
CAS-Nummer: 944692-41-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR