6-Bromo-8-chloro-2-oxo-2H-chromene-3-carboxylic acid
Lieferant: Apollo Scientific
|
Achtung
|
OR54702-100MG
OR54702-1G
OR54702-250MG
APOSOR54702-100MGEA
116
EUR
APOSOR54702-100MG
APOSOR54702-1G
APOSOR54702-250MG
6-Bromo-8-chloro-2-oxo-2H-chromene-3-carboxylic acid
6-Bromo-8-chloro-2-oxo-2H-chromene-3-carboxylic acid
Formel:
C₁₀H₄BrClO₄ Molecular Weight: 303.493 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD32177105 CAS-Nummer: 1243538-50-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...