2-Amino-N-benzyl-3-methoxypropionamide

Lieferant: Apollo Scientific

OR54708-1G OR54708-250MG OR54708-5G
APOSOR54708-1GEA 143 EUR
APOSOR54708-1G APOSOR54708-250MG APOSOR54708-5G
2-Amino-N-benzyl-3-methoxypropionamide
2-Amino-N-benzyl-3-methoxypropionamide
Formel: C₁₁H₁₆N₂O₂
Molecular Weight: 208.257 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20486464
CAS-Nummer: 262845-82-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR