7-Oxa-6-thia-5-azaspiro[3.4]octane 6,6-dioxide, N-BOC protected
Lieferant: Apollo Scientific
Achtung
Synonyme: tert-Butyl 7-oxa-6-thia-5-azaspiro[3.4]octane-5-carboxylate 6,6-dioxide
OR54830-1G OR54830-250MG OR54830-5G
APOSOR54830-1GEA 676 EUR
APOSOR54830-1G APOSOR54830-250MG APOSOR54830-5G
7-Oxa-6-thia-5-azaspiro[3.4]octane 6,6-dioxide, N-BOC protected
7-Oxa-6-thia-5-azaspiro[3.4]octane 6,6-dioxide, N-BOC protected
Formel: C₁₀H₁₇NO₅S
Molecular Weight: 263.311 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD31743876
CAS-Nummer: 2227204-96-6