(S)-2-Amino-3-methoxypropanoic acid 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: O-Methyl-L-serine

OR55011-10G OR55011-1G OR55011-5G
APOSOR55011-10GEA 616 EUR
APOSOR55011-10G APOSOR55011-1G APOSOR55011-5G
(S)-2-Amino-3-methoxypropanoic acid 97%
(S)-2-Amino-3-methoxypropanoic acid
Formel: C₄H₉NO₃
Molecular Weight: 119,119 g/mol
Schmelzpunkt: 210…215 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00145257
CAS-Nummer: 32620-11-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR