Hexadecamethylheptasiloxane

Lieferant: Apollo Scientific
Achtung

OR55032-1G OR55032-250MG
APOSOR55032-1GEA 126 EUR
APOSOR55032-1G APOSOR55032-250MG
Hexadecamethylheptasiloxane
Hexadecamethylheptasiloxane
Formel: C₁₆H₄₈O₆Si₇
Lagertemperatur: Raumtemperatur
MDL: MFCD05663924
CAS-Nummer: 541-01-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR