3-Amino-4-bromobenzonitrile
Lieferant: Apollo Scientific
|
Achtung
|
OR55073-1G
OR55073-250MG
APOSOR55073-1GEA
161
EUR
APOSOR55073-1G
APOSOR55073-250MG
3-Amino-4-bromobenzonitrile
3-Amino-4-bromobenzonitrile
Formel:
C₇H₅BrN₂ Molecular Weight: 197.032 g/mol Schmelzpunkt: 99 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD11036380 CAS-Nummer: 72635-78-0 UN: 3439 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...