5-Bromo-6-methoxypyridine-3-carboxylic acid
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-Bromo-6-methoxynicotinic acid
OR55103-100MG
OR55103-250MG
APOSOR55103-100MGEA
21.9
EUR
APOSOR55103-100MG
APOSOR55103-250MG
5-Bromo-6-methoxypyridine-3-carboxylic acid
5-Bromo-6-methoxypyridine-3-carboxylic acid
Formel:
C₇H₆BrNO₃ Molecular Weight: 232,031 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11226508 CAS-Nummer: 1186194-46-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...