6-Iodo-1H-indazole-3-carboxylic acid 97%

Lieferant: Apollo Scientific
Achtung

OR55105-100MG
APOSOR55105-100MGEA 60.9 EUR
APOSOR55105-100MG
6-Iodo-1H-indazole-3-carboxylic acid 97%
6-Iodo-1H-indazole-3-carboxylic acid
Formel: C₈H₅IN₂O₂
Molecular Weight: 288,042 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07378935
CAS-Nummer: 885520-67-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR