Pyrrolidine-3-carbonitrile 96%

Lieferant: Apollo Scientific
Achtung

OR55107-1G OR55107-250MG
APOSOR55107-1GEA 473 EUR
APOSOR55107-1G APOSOR55107-250MG
Pyrrolidine-3-carbonitrile 96%
Pyrrolidine-3-carbonitrile
Formel: C₅H₈N₂
Molecular Weight: 96,13 g/mol
Siedepunkt: 208 °C (760 mm Hg)
Lagertemperatur: Raumtemperatur
MDL: MFCD06658510
CAS-Nummer: 10603-53-9
UN: 3276
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR