5-Bromo-1,3-benzoxazole-2-carboxylic acid 98%

Lieferant: Apollo Scientific

OR55114-1G OR55114-250MG
APOSOR55114-1GEA 0 EUR
APOSOR55114-1G APOSOR55114-250MG
5-Bromo-1,3-benzoxazole-2-carboxylic acid 98%
5-Bromo-1,3-benzoxazole-2-carboxylic acid
Formel: C₈H₄BrNO₃
Molecular Weight: 242,026 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09832201
CAS-Nummer: 944898-52-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR