4-Bromo-1-cyclobutylpyrazole 95%

Lieferant: Apollo Scientific
Achtung

OR55138-1G OR55138-250MG OR55138-5G
APOSOR55138-1GEA 284 EUR
APOSOR55138-1G APOSOR55138-250MG APOSOR55138-5G
4-Bromo-1-cyclobutylpyrazole 95%
4-Bromo-1-cyclobutylpyrazole
Formel: C₇H₉BrN₂
Molecular Weight: 201.064 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD2110075
CAS-Nummer: 1002309-50-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR