Methyl 3-formyl-2-nitrobenzoate 99%
Lieferant: Apollo Scientific
|
Achtung
|
OR55158-1G
OR55158-5G
APOSOR55158-1GEA
34.8
EUR
APOSOR55158-1G
APOSOR55158-5G
Methyl 3-formyl-2-nitrobenzoate 99%
Methyl 3-formyl-2-nitrobenzoate
Formel:
C₉H₇NO₅ Molecular Weight: 209,156 g/mol Siedepunkt: 354 °C Flammpunkt: 173 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD14706049 CAS-Nummer: 138229-59-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...