4-(Methoxymethyl)piperidine

Lieferant: Apollo Scientific
Achtung

OR55162-1G OR55162-5G
APOSOR55162-1GEA 55.1 EUR
APOSOR55162-1G APOSOR55162-5G
4-(Methoxymethyl)piperidine
4-(Methoxymethyl)piperidine
Formel: C₇H₁₅NO
Molecular Weight: 129,2 g/mol
Siedepunkt: 80 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06248041
CAS-Nummer: 399580-55-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR