8-Aminoquinoline-7-carboxaldehyde
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
8-Aminoquinoline-7-carbaldehyde
OR55179-1G
OR55179-250MG
APOSOR55179-1GEA
296
EUR
APOSOR55179-1G
APOSOR55179-250MG
8-Aminoquinoline-7-carboxaldehyde
8-Aminoquinoline-7-carboxaldehyde
Formel:
C₁₀H₈N₂O Molecular Weight: 172,183 g/mol Schmelzpunkt: 85…86 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD11110554 CAS-Nummer: 158753-17-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...