4-Chloro-2-(methylthio)-5-pyrimidinecarbonyl chloride 95%
Lieferant: Apollo Scientific
Synonyme:
4-Chloro-2-(methylthio)pyrimidine-5-carbonyl chloride
OR55187-5G
APOSOR55187-5GEA
190
EUR
APOSOR55187-5G
4-Chloro-2-(methylthio)-5-pyrimidinecarbonyl chloride 95%
4-Chloro-2-(methylthio)-5-pyrimidinecarbonyl chloride
Formel:
C₆H₄Cl₂N₂OS Molecular Weight: 223,08 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD12545984 CAS-Nummer: 55084-66-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...