(1S,4S)-2-Methyl-2,5-diazabicyclo[2.2.1]heptane dihydrobromide

Lieferant: Apollo Scientific
Achtung

OR55209-1G OR55209-5G
APOSOR55209-1GEA 282 EUR
APOSOR55209-1G APOSOR55209-5G
(1S,4S)-2-Methyl-2,5-diazabicyclo[2.2.1]heptane dihydrobromide
(1S,4S)-2-Methyl-2,5-diazabicyclo[2.2.1]heptane dihydrobromide
Formel: C₆H₁₂N₂·2BrH
Molecular Weight: 273,997 g/mol
Schmelzpunkt: 220…225 °C
Lagertemperatur: Kühlschrank
MDL: MFCD01863510
CAS-Nummer: 125224-62-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR