3-Formyl-2-nitrobenzoic acid

Lieferant: Apollo Scientific
Achtung

OR55237-250MG OR55237-1G
APOSOR55237-250MGEA 169 EUR
APOSOR55237-250MG APOSOR55237-1G
3-Formyl-2-nitrobenzoic acid
3-Formyl-2-nitrobenzoic acid
Formel: C₈H₅NO₅
Molecular Weight: 195.129 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD26855790
CAS-Nummer: 1289174-44-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR