2-Chlorodiphenylamine

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Chloro-N-phenylaniline

OR55250-1G OR55250-5G
APOSOR55250-1GEA 132 EUR
APOSOR55250-1G APOSOR55250-5G
2-Chlorodiphenylamine
2-Chlorodiphenylamine
Formel: C₁₂H₁₀ClN
Molecular Weight: 203,668 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01027199
CAS-Nummer: 1205-40-9
UN: 3077
ADR: 9+EHS,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR