3-Bromo-4-chloro-2-nitroaniline

Lieferant: Apollo Scientific
Achtung

OR55252-1G OR55252-250MG OR55252-5G
APOSOR55252-1GEA 1440 EUR
APOSOR55252-1G APOSOR55252-250MG APOSOR55252-5G
3-Bromo-4-chloro-2-nitroaniline
3-Bromo-4-chloro-2-nitroaniline
Formel: C₆H₄BrClN₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD31543878
CAS-Nummer: 2091408-21-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR