2-Bromo-3-methoxybenzonitrile 97%
Lieferant: Apollo Scientific
|
Achtung
|
OR55274-250MG
OR55274-1G
OR55274-5G
APOSOR55274-250MGEA
97.3
EUR
APOSOR55274-250MG
APOSOR55274-1G
APOSOR55274-5G
2-Bromo-3-methoxybenzonitrile 97%
2-Bromo-3-methoxybenzonitrile
Formel:
C₈H₆BrNO Molecular Weight: 212,043 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09834772 CAS-Nummer: 1261816-95-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...