2-Methoxy-3-methylbenzonitrile 97%

Lieferant: Apollo Scientific
Achtung

OR55275-250MG OR55275-1G
APOSOR55275-250MGEA 174 EUR
APOSOR55275-250MG APOSOR55275-1G
2-Methoxy-3-methylbenzonitrile 97%
2-Methoxy-3-methylbenzonitrile
Formel: C₉H₉NO
Molecular Weight: 147,174 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13704701
CAS-Nummer: 53078-68-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR