3,5-Diethynylbenzoic acid

Lieferant: Apollo Scientific
Achtung

OR55280-250MG OR55280-1G
APOSOR55280-250MGEA 267 EUR
APOSOR55280-250MG APOSOR55280-1G
3,5-Diethynylbenzoic acid
3,5-Diethynylbenzoic acid
Formel: C₁₁H₆O₂
Lagertemperatur: Kühlschrank
MDL: MFCD06658429
CAS-Nummer: 883106-26-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR