4-(Methoxycarbonyl)benzeneboronic acid pinacol ester

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl 4-((4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)methyl)benzoate, Methyl 4-[(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)methyl]benzoate

OR55281-1G OR55281-250MG
APOSOR55281-1GEA 162 EUR
APOSOR55281-1G APOSOR55281-250MG
4-(Methoxycarbonyl)benzeneboronic acid pinacol ester
4-(Methoxycarbonyl)benzeneboronic acid pinacol ester
Formel: C₁₅H₂₁BO₄
Molecular Weight: 276.136 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD18383373
CAS-Nummer: 150033-80-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR