trans-Ethyl 4-aminocyclohexanecarboxylate hydrochloride
Lieferant: Apollo Scientific
Synonyme:
Ethyl 4-aminocyclohexane-1-carboxylate, hydrochloride
OR55285-250MG
OR55285-1G
APOSOR55285-250MGEA
50.9
EUR
APOSOR55285-250MG
APOSOR55285-1G
trans-Ethyl 4-aminocyclohexanecarboxylate hydrochloride
trans-Ethyl 4-aminocyclohexanecarboxylate hydrochloride
Formel:
C₉H₁₇NO₂·ClH Molecular Weight: 207,698 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD14635812 CAS-Nummer: 2084-28-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...