Iodocholine iodide 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-hydroxyethyl-(iodomethyl)-dimethylazanium iodide

OR55305-1G OR55305-100MG
APOSOR55305-1GEA 480 EUR
APOSOR55305-1G APOSOR55305-100MG
Iodocholine iodide 97%
Iodocholine iodide
Formel: C₅H₁₃INO·I
Molecular Weight: 356,972 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20489393
CAS-Nummer: 28508-22-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR