5-Amino-2,4,6-trichloropyrimidine 90%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
2,4,6-Trichloropyrimidin-5-amine
OR55310-250MG
OR55310-1G
APOSOR55310-250MGEA
125
EUR
APOSOR55310-250MG
APOSOR55310-1G
5-Amino-2,4,6-trichloropyrimidine 90%
5-Amino-2,4,6-trichloropyrimidine
Formel:
C₄H₂Cl₃N₃ Molecular Weight: 198,438 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08693695 CAS-Nummer: 91322-00-8 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...