Methyl 2-[6-(chloromethyl)-2-pyridyl]acetate hydrochloride

Lieferant: Apollo Scientific
Gefahr

OR55327-250MG OR55327-1G OR55327-100MG
APOSOR55327-250MGEA 481 EUR
APOSOR55327-250MG APOSOR55327-1G APOSOR55327-100MG
Methyl 2-[6-(chloromethyl)-2-pyridyl]acetate hydrochloride
Methyl 2-[6-(chloromethyl)-2-pyridyl]acetate hydrochloride
Formel: C₉H₁₀ClNO₂·ClH
Molecular Weight: 236,095 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31619564
CAS-Nummer: 2227272-88-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR