5-Bromo-6-chloro-isatoic anhydride 95%

Lieferant: Apollo Scientific
Achtung

OR55334-250MG OR55334-1G
APOSOR55334-250MGEA 219 EUR
APOSOR55334-250MG APOSOR55334-1G
5-Bromo-6-chloro-isatoic anhydride 95%
5-Bromo-6-chloro-isatoic anhydride
Formel: C₈H₃BrClNO₃
Molecular Weight: 276.471 g/mol
Schmelzpunkt: 273 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD31002307
CAS-Nummer: 943138-45-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR