4'-Bromo-3'-iodoacetophenone 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-(4-Bromo-3-iodophenyl)ethanone

OR55348-250MG OR55348-1G OR55348-5G
APOSOR55348-250MGEA 21.9 EUR
APOSOR55348-250MG APOSOR55348-1G APOSOR55348-5G
4'-Bromo-3'-iodoacetophenone 95%
4'-Bromo-3'-iodoacetophenone
Formel: C₈H₆BrIO
Molecular Weight: 324,941 g/mol
Schmelzpunkt: 79…82 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD18391612
CAS-Nummer: 919124-08-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR