5-Iodobenzo[b]furan

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Iodo-1-benzofuran, 5-Iodobenzofuran

OR55385-250MG OR55385-1G OR55385-100MG
APOSOR55385-250MGEA 72.5 EUR
APOSOR55385-250MG APOSOR55385-1G APOSOR55385-100MG
5-Iodobenzo[b]furan
5-Iodobenzo[b]furan
Formel: C₈H₅IO
Molecular Weight: 244,029 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12406296
CAS-Nummer: 60770-67-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR