(2R,5S)-2,5-Dimethylpiperazine, N1-BOC protected 95+%

Lieferant: Apollo Scientific
Achtung

Synonyme: (2R,5S)-tert-Butyl 2,5-dimethylpiperazine-1-carboxylate, tert-butyl (2R,5S)-2,5-dimethylpiperazine-1-carboxylate

OR55395-5G
APOSOR55395-5GEA 55.1 EUR
APOSOR55395-5G
(2R,5S)-2,5-Dimethylpiperazine, N1-BOC protected 95+%
(2R,5S)-2,5-Dimethylpiperazine, N1-BOC protected
Formel: C₁₁H₂₂N₂O₂
Molecular Weight: 214,305 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD06797728
CAS-Nummer: 309915-46-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR