2-Amino-6-nitrobenzyl alcohol
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(2-Amino-6-nitrophenyl)methanol
OR55410-1G
OR55410-5G
OR55410-10G
APOSOR55410-1GEA
132
EUR
APOSOR55410-1G
APOSOR55410-5G
APOSOR55410-10G
2-Amino-6-nitrobenzyl alcohol
2-Amino-6-nitrobenzyl alcohol
Formel:
C₇H₈N₂O₃ Molecular Weight: 168,15 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01664411 CAS-Nummer: 98451-51-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...