4-Bromo-3-iodo-2-nitroaniline 95%
Lieferant: Apollo Scientific
|
Achtung
|
OR55422-1G
OR55422-250MG
APOSOR55422-1GEA
654
EUR
APOSOR55422-1G
APOSOR55422-250MG
4-Bromo-3-iodo-2-nitroaniline 95%
4-Bromo-3-iodo-2-nitroaniline
Formel:
C₆H₄BrIN₂O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD31619569 CAS-Nummer: 2091791-61-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...